missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzohydrazide 98.0+%, TCI America™

Catalog No. F05405G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05405G 5 g
F054025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05405G Supplier TCI America Supplier No. F05405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 456-06-4
Molecular Formula C7H7FN2O
Molecular Weight (g/mol) 154.144
InChI Key UIVXXFYJRYVRKJ-UHFFFAOYSA-N
Synonym 4-fluorobenzhydrazide, 4-fluorobenzoic hydrazide, p-fluorobenzoic acid hydrazide, p-fluorobenzoylhydrazine, 4-fluorobenzoic acid hydrazide, hydrazine, p-fluorobenzoyl, 4-fluoro benzoyl hydrazine, benzoic acid, p-fluoro-, hydrazide, 4-fluorobenzenecarbohydrazide, 4-fluoro-benzoic acid hydrazide
PubChem CID 9972
IUPAC Name 4-fluorobenzohydrazide
SMILES C1=CC(=CC=C1C(=O)NN)F

Specifications

Melting Point 165°C
Color White-Yellow
Quantity 5 g
Formula Weight 154.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorobenzohydrazide
RTECSNumber DH1310000
TSCA No