missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzenethiol 98.0+%, TCI America™

Supplier:  TCI America F039325G

 View more versions of this product

Catalog No. F039325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Fluorobenzenethiol
Yellow
C6H5FS
25 g
4-fluorothiophenol, p-fluorothiophenol, 4-fluorobenzene-1-thiol, 4-fluoro thiophenol, p-fluorophenyl mercaptan, benzenethiol, 4-fluoro, 4-fluoro-benzenethiol, 4-mercaptofluorobenzene, 4-fluorthiophenol, p-fluoro-thiophenol
C1=CC(=CC=C1F)S
128.164
128.16
Liquid
371-42-6
168°C
MFCD00004846
3336
OKIHXNKYYGUVTE-UHFFFAOYSA-N
4-fluorobenzenethiol
67789
≥98.0% (GC)

Chemical Identifiers

371-42-6
128.164
OKIHXNKYYGUVTE-UHFFFAOYSA-N
67789
C1=CC(=CC=C1F)S
C6H5FS
MFCD00004846
4-fluorothiophenol, p-fluorothiophenol, 4-fluorobenzene-1-thiol, 4-fluoro thiophenol, p-fluorophenyl mercaptan, benzenethiol, 4-fluoro, 4-fluoro-benzenethiol, 4-mercaptofluorobenzene, 4-fluorthiophenol, p-fluoro-thiophenol
4-fluorobenzenethiol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions