missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzenethiol 98.0+%, TCI America™

Catalog No. F03935G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03935G 5 g
F039325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03935G Supplier TCI America Supplier No. F03935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 371-42-6
Molecular Formula C6H5FS
Molecular Weight (g/mol) 128.164
MDL Number MFCD00004846
InChI Key OKIHXNKYYGUVTE-UHFFFAOYSA-N
Synonym 4-fluorothiophenol, p-fluorothiophenol, 4-fluorobenzene-1-thiol, 4-fluoro thiophenol, p-fluorophenyl mercaptan, benzenethiol, 4-fluoro, 4-fluoro-benzenethiol, 4-mercaptofluorobenzene, 4-fluorthiophenol, p-fluoro-thiophenol
PubChem CID 67789
IUPAC Name 4-fluorobenzenethiol
SMILES C1=CC(=CC=C1F)S

Specifications

Color Yellow
Boiling Point 168°C
Quantity 5 g
UN Number 3336
Formula Weight 128.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluorobenzenethiol