Learn More
4-Fluorobenzaldehyde, 98%
CAS: 459-57-4 | C7H5FO | 124.114 g/mol
Supplier: Thermo Scientific Chemicals A1538322
Description
4-Fluorobenzaldehyde is used in the preparation of pyrazolopyridine derivative, which finds application as mitogen-activated protein kinase (MAPK) inhibitor. It also serves as a synthetic intermediate in the preparation of pharmaceutical compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluorobenzaldehyde is used in the preparation of pyrazolopyridine derivative, which finds application as mitogen-activated protein kinase (MAPK) inhibitor. It also serves as a synthetic intermediate in the preparation of pharmaceutical compounds.
Solubility
Immiscible with water.
Notes
Air sensitive. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
Specifications
| 4-Fluorobenzaldehyde | |
| -10°C | |
| 180°C to 181°C | |
| C7H5FO | |
| MFCD00003378 | |
| UN1989 | |
| Air Sensitive | |
| Immiscible with water. | |
| C1=CC(=CC=C1C=O)F | |
| 124.114 | |
| 124.11 |
| 459-57-4 | |
| 1.175 | |
| 56°C (133°F) | |
| 1.521 | |
| 100 g | |
| 385857 | |
| p-fluorobenzaldehyde, benzaldehyde, 4-fluoro, 4-fluoro-benzaldehyde, para-fluorobenzaldehyde, p-fluorbenzaldehyde, fluorobenzaldehyde 4-, benzaldehyde, p-fluoro, pfad, 4-fluoro benzaldehyde, para fluoro benzaldehyde | |
| UOQXIWFBQSVDPP-UHFFFAOYSA-N | |
| 4-fluorobenzaldehyde | |
| 68023 | |
| 98% |
Chemical Identifiers
| 459-57-4 | |
| 124.114 | |
| UOQXIWFBQSVDPP-UHFFFAOYSA-N | |
| 68023 | |
| C1=CC(=CC=C1C=O)F |
| C7H5FO | |
| MFCD00003378 | |
| p-fluorobenzaldehyde, benzaldehyde, 4-fluoro, 4-fluoro-benzaldehyde, para-fluorobenzaldehyde, p-fluorbenzaldehyde, fluorobenzaldehyde 4-, benzaldehyde, p-fluoro, pfad, 4-fluoro benzaldehyde, para fluoro benzaldehyde | |
| 4-fluorobenzaldehyde |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.
EINECSNumber : 207-293-6
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only