missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzaldehyde, 98+%

Catalog No. AC156152500
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156152500 250 mL Glass bottle
AC156150500 50 mL Glass bottle
AC156150010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC156152500 Supplier Thermo Scientific Chemicals Supplier No. 156152500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 459-57-4 | C7H5FO | 124.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 459-57-4
Molecular Formula C7H5FO
Molecular Weight (g/mol) 124.11
MDL Number MFCD00003378
InChI Key UOQXIWFBQSVDPP-UHFFFAOYSA-N
Synonym p-fluorobenzaldehyde, benzaldehyde, 4-fluoro, 4-fluoro-benzaldehyde, para-fluorobenzaldehyde, p-fluorbenzaldehyde, fluorobenzaldehyde 4-, benzaldehyde, p-fluoro, pfad, 4-fluoro benzaldehyde, para fluoro benzaldehyde
PubChem CID 68023
IUPAC Name 4-fluorobenzaldehyde
SMILES C1=CC(=CC=C1C=O)F

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -10.0°C
Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 181.0°C (758.0 mmHg)
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5195 to 1.5215
Linear Formula FC6H4CHO
Quantity 250 mL
Beilstein 07, I, 132
Specific Gravity 1.15
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in ether,acetone,benzene and alcohol
Formula Weight 124.11
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 4-Fluorobenzaldehyde, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-293-6
TSCA TSCA

RUO – Research Use Only