missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoroanisole 97.0+%, TCI America™

Catalog No. p-7129557
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
F023125G 25 g
F0231500G 500 g
2 options

Catalog No. F023125G

Supplier: TCI America F023125G

Only null left

Chemical Identifiers

CAS 459-60-9
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00000348
InChI Key VIPWUFMFHBIKQI-UHFFFAOYSA-N
Synonym 4-fluoroanisole, p-fluoroanisole, benzene, 1-fluoro-4-methoxy, p-fluoromethoxybenzene, p-methoxyfluorobenzene, 4-fluoroanisol, p-fluorophenyl methyl ether, anisole, p-fluoro, 1-fluoro-4-methoxy-benzene, 4-methoxyfluorobenzene
PubChem CID 9987
IUPAC Name 1-fluoro-4-methoxybenzene
SMILES COC1=CC=C(C=C1)F

Specifications

Color Yellow
Boiling Point 157°C
Quantity 25 g
UN Number 1993
Formula Weight 126.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluoroanisole