Learn More
4-Fluoroaniline, 99%
CAS: 371-40-4 | C6H6FN | 111.12 g/mol
Supplier: Thermo Scientific Chemicals A1559722
Description
It finds its application as an analytical reagent in the enzymic detection of glucose. It is used as an intermediate in the manufacture of pharmaceuticals and plant growth regulators. It is employed as an intermediate for the production of Flumazenil.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its application as an analytical reagent in the enzymic detection of glucose. It is used as an intermediate in the manufacture of pharmaceuticals and plant growth regulators. It is employed as an intermediate for the production of Flumazenil.
Solubility
Slightly soluble in water. Soluble in chloroform and methanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 4-Fluoroaniline | |
| -2°C | |
| 186°C to 187°C | |
| Recognizable | |
| 1.54 | |
| 100 g | |
| 742030 | |
| p-fluoroaniline, benzenamine, 4-fluoro, 1-amino-4-fluorobenzene, 4-fluorobenzenamine, p-fluorophenylamine, 4-fluoronaniline, aniline, p-fluoro, aniline, 4-fluoro, 4-fluoranilin, para-fluoroaniline | |
| KRZCOLNOCZKSDF-UHFFFAOYSA-N | |
| 4-fluoroaniline | |
| 9731 | |
| 111.12 |
| 371-40-4 | |
| 1.158 | |
| 73°C (163°F) | |
| C6H6FN | |
| MFCD00007829 | |
| UN2941 | |
| 14,4169 | |
| Slightly soluble in water. Soluble in chloroform and methanol. | |
| NC1=CC=C(F)C=C1 | |
| 111.12 | |
| CHEBI:28546 | |
| 99% |
Chemical Identifiers
| 371-40-4 | |
| 111.12 | |
| KRZCOLNOCZKSDF-UHFFFAOYSA-N | |
| 9731 | |
| 4-fluoroaniline |
| C6H6FN | |
| MFCD00007829 | |
| p-fluoroaniline, benzenamine, 4-fluoro, 1-amino-4-fluorobenzene, 4-fluorobenzenamine, p-fluorophenylamine, 4-fluoronaniline, aniline, p-fluoro, aniline, 4-fluoro, 4-fluoranilin, para-fluoroaniline | |
| CHEBI:28546 | |
| NC1=CC=C(F)C=C1 |
Safety and Handling
GHS H Statement
H331-H314-H318-H373-H227-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302-H314-H335-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: FLUOROANILINES
EINECSNumber : 206-735-5
RTECSNumber : BY1575000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only