missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoroaniline, 98%

Catalog No. p-134242
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC119290250 25 mL Glass bottle
AC119291000 100 mL Glass bottle
AC119295000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC119290250 Supplier Thermo Scientific Chemicals Supplier No. 119290250

CAS: 371-40-4 | C6H6FN | 111.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 371-40-4
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.12
MDL Number MFCD00007829
InChI Key KRZCOLNOCZKSDF-UHFFFAOYSA-N
Synonym p-fluoroaniline, benzenamine, 4-fluoro, 1-amino-4-fluorobenzene, 4-fluorobenzenamine, p-fluorophenylamine, 4-fluoronaniline, aniline, p-fluoro, aniline, 4-fluoro, 4-fluoranilin, para-fluoroaniline
PubChem CID 9731
ChEBI CHEBI:28546
IUPAC Name 4-fluoroaniline
SMILES NC1=CC=C(F)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -1.9°C
Density 1.1570g/mL
Boiling Point 187.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5385 to 1.5405
Linear Formula FC6H4NH2
Quantity 25 mL
Beilstein 12, 597
Specific Gravity 1.157
Merck Index 15, 4200
Solubility Information Solubility in water: 33g/L (20°C). Other solubilities: soluble in acetone,ethanol,xylene alcohol,,carbon tetrachloride and diethyl ether
Formula Weight 111.12
Percent Purity 98%
Physical Form Oily Liquid
Chemical Name or Material 4-Fluoroaniline, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 206-735-5
RTECSNumber BY1575000
TSCA TSCA

RUO – Research Use Only