missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoroaniline 98.0+%, TCI America™
$93.21 - $93.21
Chemical Identifiers
| CAS | 371-40-4 |
|---|---|
| Molecular Formula | C6H6FN |
| Molecular Weight (g/mol) | 111.12 |
| MDL Number | MFCD00007829 |
| InChI Key | KRZCOLNOCZKSDF-UHFFFAOYSA-N |
| Synonym | p-fluoroaniline, benzenamine, 4-fluoro, 1-amino-4-fluorobenzene, 4-fluorobenzenamine, p-fluorophenylamine, 4-fluoronaniline, aniline, p-fluoro, aniline, 4-fluoro, 4-fluoranilin, para-fluoroaniline |
| PubChem CID | 9731 |
| ChEBI | CHEBI:28546 |
| IUPAC Name | 4-fluoroaniline |
| SMILES | NC1=CC=C(F)C=C1 |
Chemical Identifiers
| 371-40-4 | |
| 111.12 | |
| KRZCOLNOCZKSDF-UHFFFAOYSA-N | |
| 9731 | |
| 4-fluoroaniline |
| C6H6FN | |
| MFCD00007829 | |
| p-fluoroaniline, benzenamine, 4-fluoro, 1-amino-4-fluorobenzene, 4-fluorobenzenamine, p-fluorophenylamine, 4-fluoronaniline, aniline, p-fluoro, aniline, 4-fluoro, 4-fluoranilin, para-fluoroaniline | |
| CHEBI:28546 | |
| NC1=CC=C(F)C=C1 |
Specifications
| 371-40-4 | |
| Yellow | |
| C6H6FN | |
| 25 mL | |
| p-fluoroaniline, benzenamine, 4-fluoro, 1-amino-4-fluorobenzene, 4-fluorobenzenamine, p-fluorophenylamine, 4-fluoronaniline, aniline, p-fluoro, aniline, 4-fluoro, 4-fluoranilin, para-fluoroaniline | |
| NC1=CC=C(F)C=C1 | |
| 111.12 | |
| CHEBI:28546 | |
| ≥98.0% (GC,T) | |
| 4-Fluoroaniline |
| -1°C | |
| 188°C | |
| MFCD00007829 | |
| 2941 | |
| KRZCOLNOCZKSDF-UHFFFAOYSA-N | |
| 4-fluoroaniline | |
| 9731 | |
| 111.12 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-0222
RTECSNumber : BY1575000
TSCA : Yes