missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-trans-beta-styrylboronic acid pinacol ester, 97%

Catalog No. AAH3057803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3057803 1 g
AAH3057806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3057803 Supplier Thermo Scientific Chemicals Supplier No. H3057803

CAS: 504433-86-7 | C14H18BFO2 | 248.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 504433-86-7
Molecular Formula C14H18BFO2
Molecular Weight (g/mol) 248.10
MDL Number MFCD12546189
InChI Key ZJRAXVMUDOVAOD-UHFFFAOYSA-N
Synonym e-2-4-fluorostyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-trans-beta-styrylboronic acid pinacol ester, 2-e-2-4-fluorophenyl ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-p-fluorostyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-e-4-fluorostyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-trans-2-4-fluorophenyl vinyl-4,4,5,5-tetramethyl 1,3,2 dioxaborolane
PubChem CID 11207378
IUPAC Name 2-[(E)-2-(4-fluorophenyl)ethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C=CC1=CC=C(F)C=C1

Specifications

Quantity 1 g
Formula Weight 248.11
Percent Purity 97%
Chemical Name or Material 4-Fluoro-trans-beta-styrylboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only