missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-o-xylene 99.0+%, TCI America™

Catalog No. F035510G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F035510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F035510G Supplier TCI America Supplier No. F035510G

Chemical Identifiers

CAS 452-64-2
Molecular Formula C8H9F
Molecular Weight (g/mol) 124.158
MDL Number MFCD00039217
InChI Key WYHBENDEZDFJNU-UHFFFAOYSA-N
Synonym 4-fluoro-o-xylene, 1,2-dimethyl-4-fluorobenzene, 3,4-dimethylfluorobenzene, 4-fluoro-1,2-xylene, 1-fluoro-3,4-dimethylbenzene, benzene, 4-fluoro-1,2-dimethyl, o-xylene, 4-fluoro, pubchem14118, acmc-1afww, 3,4-dimethyl fluorobenzene
PubChem CID 253306
IUPAC Name 4-fluoro-1,2-dimethylbenzene
SMILES CC1=C(C=C(C=C1)F)C

Specifications

Color Colorless
Quantity 10 g
Physical Form Clear Liquid at 20°C
UN Number 1993
Boiling Point 60°C
Formula Weight 124.16
Percent Purity ≥99.0% (GC)
Chemical Name or Material 4-Fluoro-o-xylene