missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-o-xylene, 98%

Catalog No. p-7024072
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1825306 5 g
AAA1825314 25 g
AAA1825322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1825306 Supplier Thermo Scientific Chemicals Supplier No. A1825306

CAS: 452-64-2 | C8H9F | 124.158 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 452-64-2
Molecular Formula C8H9F
Molecular Weight (g/mol) 124.158
MDL Number MFCD00039217
InChI Key WYHBENDEZDFJNU-UHFFFAOYSA-N
Synonym 4-fluoro-o-xylene, 1,2-dimethyl-4-fluorobenzene, 3,4-dimethylfluorobenzene, 4-fluoro-1,2-xylene, 1-fluoro-3,4-dimethylbenzene, benzene, 4-fluoro-1,2-dimethyl, o-xylene, 4-fluoro, pubchem14118, acmc-1afww, 3,4-dimethyl fluorobenzene
PubChem CID 253306
IUPAC Name 4-fluoro-1,2-dimethylbenzene
SMILES CC1=C(C=C(C=C1)F)C

Specifications

Density 1
Quantity 5 g
Odor Aromatic
Refractive Index 1.4815
UN Number UN1993
Beilstein 1856457
Boiling Point 148°C to 149°C
Formula Weight 124.16
Percent Purity 98%
Chemical Name or Material 4-Fluoro-o-xylene
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only