missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-o-cresol 98.0+%, TCI America™
$87.60 - $87.60
Chemical Identifiers
| CAS | 452-72-2 |
|---|---|
| Molecular Formula | C7H7FO |
| Molecular Weight (g/mol) | 126.13 |
| MDL Number | MFCD00075088 |
| InChI Key | GKQDDKKGDIVDAG-UHFFFAOYSA-N |
| Synonym | 2-methyl-4-fluorophenol, 4-fluoro-o-cresol, 4-fluoro-2-methyl-phenol, 5-fluoro-2-hydroxytoluene, phenol, 4-fluoro-2-methyl, pubchem1500, acmc-1csjj, 4-fuoro-2-methyl-phenol, 2-methyl-4-fluoro phenol, 4-fluoro-2-methyl phenol |
| PubChem CID | 136295 |
| IUPAC Name | 4-fluoro-2-methylphenol |
| SMILES | CC1=C(C=CC(=C1)F)O |
Chemical Identifiers
| 452-72-2 | |
| 126.13 | |
| GKQDDKKGDIVDAG-UHFFFAOYSA-N | |
| 136295 | |
| CC1=C(C=CC(=C1)F)O |
| C7H7FO | |
| MFCD00075088 | |
| 2-methyl-4-fluorophenol, 4-fluoro-o-cresol, 4-fluoro-2-methyl-phenol, 5-fluoro-2-hydroxytoluene, phenol, 4-fluoro-2-methyl, pubchem1500, acmc-1csjj, 4-fuoro-2-methyl-phenol, 2-methyl-4-fluoro phenol, 4-fluoro-2-methyl phenol | |
| 4-fluoro-2-methylphenol |
Specifications
| 452-72-2 | |
| Yellow | |
| C7H7FO | |
| 5 g | |
| GKQDDKKGDIVDAG-UHFFFAOYSA-N | |
| 4-fluoro-2-methylphenol | |
| 136295 | |
| ≥98.0% (GC) | |
| 4-Fluoro-o-cresol |
| 36°C | |
| 93°C | |
| MFCD00075088 | |
| 2-methyl-4-fluorophenol, 4-fluoro-o-cresol, 4-fluoro-2-methyl-phenol, 5-fluoro-2-hydroxytoluene, phenol, 4-fluoro-2-methyl, pubchem1500, acmc-1csjj, 4-fuoro-2-methyl-phenol, 2-methyl-4-fluoro phenol, 4-fluoro-2-methyl phenol | |
| CC1=C(C=CC(=C1)F)O | |
| 126.13 | |
| 126.13 | |
| Crystals |
Safety and Handling
TSCA : No