missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-o-cresol 98.0+%, TCI America™

Catalog No. F03875G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F03875G Supplier TCI America Supplier No. F03875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 452-72-2
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00075088
InChI Key GKQDDKKGDIVDAG-UHFFFAOYSA-N
Synonym 2-methyl-4-fluorophenol, 4-fluoro-o-cresol, 4-fluoro-2-methyl-phenol, 5-fluoro-2-hydroxytoluene, phenol, 4-fluoro-2-methyl, pubchem1500, acmc-1csjj, 4-fuoro-2-methyl-phenol, 2-methyl-4-fluoro phenol, 4-fluoro-2-methyl phenol
PubChem CID 136295
IUPAC Name 4-fluoro-2-methylphenol
SMILES CC1=C(C=CC(=C1)F)O

Specifications

Melting Point 36°C
Color Yellow
Boiling Point 93°C
Quantity 5 g
Formula Weight 126.13
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 4-Fluoro-o-cresol