missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-o-anisaldehyde 97.0+%, TCI America™

Catalog No. p-7129923
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08271G 1 g
F08275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08271G Supplier TCI America Supplier No. F08271G
Only null left

Chemical Identifiers

CAS 450-83-9
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00143318
InChI Key PTKRQIRPNNIORO-UHFFFAOYSA-N
Synonym 2-methoxy-4-fluorobenzaldehyde, benzaldehyde, 4-fluoro-2-methoxy, 4-fluoro-2-methoxy-benzaldehyde, 4-fluoro-o-anisaldehyde, fluoromethoxybenzaldehyde4, pubchem2635, 5-fluoro-2-formylanisole, acmc-1ags2, ksc494s3t, 4-fluoro-2-methoxybenzal dehyde
PubChem CID 2774537
IUPAC Name 4-fluoro-2-methoxybenzaldehyde
SMILES COC1=CC(F)=CC=C1C=O

Specifications

Melting Point 61°C
Color White-Yellow
Quantity 1 g
Formula Weight 154.14
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Fluoro-o-anisaldehyde