missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-N,N-dimethylaniline 98.0+%, TCI America™

Catalog No. p-7129522
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F01751G 1 g
F01755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F01751G Supplier TCI America Supplier No. F01751G

Chemical Identifiers

CAS 403-46-3
Molecular Formula C8H10FN
Molecular Weight (g/mol) 139.173
MDL Number MFCD00059471
InChI Key YJEHCGOJNJUOII-UHFFFAOYSA-N
PubChem CID 136249
IUPAC Name 4-fluoro-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)F

Specifications

Melting Point 36°C
Color White-Yellow
Boiling Point 200°C
Quantity 1 g
Formula Weight 139.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Fluoro-N,N-dimethylaniline