accessibility menu, dialog, popup

UserName

CAS RN 403-46-3

IUPAC Name: 4-fluoro-N,N-dimethylaniline
Synonyms: 4-fluoro-N,N-dimethylaniline
Molecular Weight (g/mol): 139.17
Molecular Formula: C8H10FN
InChi Key: YJEHCGOJNJUOII-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results