missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-N-(4-methylphenyl)benzamide, 97%, Thermo Scientific™

Catalog No. aah5719103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5719103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5719103 Supplier Thermo Scientific Chemicals Supplier No. H5719103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 399-05-3
Molecular Formula C14H12FNO
Molecular Weight (g/mol) 229.254
MDL Number MFCD00446355
InChI Key FYIMLKPLYFXRDM-UHFFFAOYSA-N
Synonym 4-fluoro-n-4-methylphenyl benzamide, 4-fluoro-n-p-tolyl-benzamide, 4-fluoro-n-p-tolyl benzamide, n-4-methylphenyl-4-fluorobenzamide, benzamide,4-fluoro-n-4-methylphenyl, 4-fluoro-n∼1∼-4-methylphenyl benzamide, 4-fluorophenyl-n-4-methylphenyl carboxamide
PubChem CID 693096
IUPAC Name 4-fluoro-N-(4-methylphenyl)benzamide
SMILES CC1=CC=C(C=C1)NC(=O)C2=CC=C(C=C2)F

Specifications

Quantity 1 g
Formula Weight 229.25
Percent Purity 97%
Chemical Name or Material 4-Fluoro-N-(4-methylphenyl)benzamide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures