missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-N-(4-fluorobenzylidene)aniline 98.0+%, TCI America™
$84.70 - $275.30
Chemical Identifiers
| CAS | 39769-09-0 |
|---|---|
| Molecular Formula | C13H9F2N |
| Molecular Weight (g/mol) | 217.219 |
| MDL Number | MFCD00017950 |
| InChI Key | FRNJXANITCYEMD-UHFFFAOYSA-N |
| Synonym | 4,4'-difluorobenzylideneaniline, 4-fluoro-n-4-fluorobenzylidene aniline, n,1-bis 4-fluorophenyl methanimine, 1e-1,2-bis 4-fluorophenyl-1-azaethene, 4-fluorobenzylidene 4-fluorophenyl amine, n-4-fluorophenyl-4-fluorobenzylideneamine, n-4-fluorobenzylidene-4-fluoroaniline, 4-fluoro-n-4-fluorophenyl methylidene aniline, 1e-n,1-bis 4-fluorophenyl methanimine |
| PubChem CID | 2734613 |
| IUPAC Name | N,1-bis(4-fluorophenyl)methanimine |
| SMILES | C1=CC(=CC=C1C=NC2=CC=C(C=C2)F)F |
Chemical Identifiers
| 39769-09-0 | |
| 217.219 | |
| FRNJXANITCYEMD-UHFFFAOYSA-N | |
| 2734613 | |
| C1=CC(=CC=C1C=NC2=CC=C(C=C2)F)F |
| C13H9F2N | |
| MFCD00017950 | |
| 4,4'-difluorobenzylideneaniline, 4-fluoro-n-4-fluorobenzylidene aniline, n,1-bis 4-fluorophenyl methanimine, 1e-1,2-bis 4-fluorophenyl-1-azaethene, 4-fluorobenzylidene 4-fluorophenyl amine, n-4-fluorophenyl-4-fluorobenzylideneamine, n-4-fluorobenzylidene-4-fluoroaniline, 4-fluoro-n-4-fluorophenyl methylidene aniline, 1e-n,1-bis 4-fluorophenyl methanimine | |
| N,1-bis(4-fluorophenyl)methanimine |
Specifications
| 39769-09-0 | |
| Yellow | |
| MFCD00017950 | |
| 4,4'-difluorobenzylideneaniline, 4-fluoro-n-4-fluorobenzylidene aniline, n,1-bis 4-fluorophenyl methanimine, 1e-1,2-bis 4-fluorophenyl-1-azaethene, 4-fluorobenzylidene 4-fluorophenyl amine, n-4-fluorophenyl-4-fluorobenzylideneamine, n-4-fluorobenzylidene-4-fluoroaniline, 4-fluoro-n-4-fluorophenyl methylidene aniline, 1e-n,1-bis 4-fluorophenyl methanimine | |
| C1=CC(=CC=C1C=NC2=CC=C(C=C2)F)F | |
| 217.219 | |
| 217.22 | |
| Crystalline Powder |
| 65°C | |
| C13H9F2N | |
| 1 g | |
| FRNJXANITCYEMD-UHFFFAOYSA-N | |
| N,1-bis(4-fluorophenyl)methanimine | |
| 2734613 | |
| ≥98.0% (GC,T) | |
| 4-Fluoro-N-(4-fluorobenzylidene)aniline |
Safety and Handling
TSCA : No