missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-N-(4-fluorobenzylidene)aniline 98.0+%, TCI America™

Catalog No. F08921G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08921G 1 g
F08925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08921G Supplier TCI America Supplier No. F08921G
Only null left

Chemical Identifiers

CAS 39769-09-0
Molecular Formula C13H9F2N
Molecular Weight (g/mol) 217.219
MDL Number MFCD00017950
InChI Key FRNJXANITCYEMD-UHFFFAOYSA-N
Synonym 4,4'-difluorobenzylideneaniline, 4-fluoro-n-4-fluorobenzylidene aniline, n,1-bis 4-fluorophenyl methanimine, 1e-1,2-bis 4-fluorophenyl-1-azaethene, 4-fluorobenzylidene 4-fluorophenyl amine, n-4-fluorophenyl-4-fluorobenzylideneamine, n-4-fluorobenzylidene-4-fluoroaniline, 4-fluoro-n-4-fluorophenyl methylidene aniline, 1e-n,1-bis 4-fluorophenyl methanimine
PubChem CID 2734613
IUPAC Name N,1-bis(4-fluorophenyl)methanimine
SMILES C1=CC(=CC=C1C=NC2=CC=C(C=C2)F)F

Specifications

Melting Point 65°C
Color Yellow
Quantity 1 g
Formula Weight 217.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-N-(4-fluorobenzylidene)aniline