missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-L-phenylalanine, 98+%
CAS: 1132-68-9 | C9H10FNO2 | 183.182 g/mol
$139.29 - $1502.27
Chemical Identifiers
| CAS | 1132-68-9 |
|---|---|
| Molecular Formula | C9H10FNO2 |
| Molecular Weight (g/mol) | 183.182 |
| MDL Number | MFCD00063064 |
| InChI Key | XWHHYOYVRVGJJY-QMMMGPOBSA-N |
| Synonym | 4-fluoro-l-phenylalanine, p-fluoro-l-phenylalanine, s-4-fluorophenylalanine, l-4-fluorophenylalanine, h-phe 4-f-oh, s-2-amino-3-4-fluorophenyl propanoic acid, unii-e2k2vdk6kk, l-phenylalanine, 4-fluoro, s---p-fluorophenylalanine, 2s-2-amino-3-4-fluorophenyl propanoic acid |
| PubChem CID | 716312 |
| ChEBI | CHEBI:44909 |
| IUPAC Name | (2S)-2-amino-3-(4-fluorophenyl)propanoic acid |
| SMILES | C1=CC(=CC=C1CC(C(=O)O)N)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL19934MD
|
Thermo Scientific Chemicals
L19934MD |
250 mg |
Each for $139.29
|
|
|||||
|
AAL1993403
|
Thermo Scientific Chemicals
L1993403 |
1 g |
Each for $397.19
|
|
|||||
|
AAL1993406
|
Thermo Scientific Chemicals
L1993406 |
5 g |
Each for $1,502.27
|
|
|||||
Description
4-Fluoro-L-phenylalanine is a substrate for tyrosine hydroxylase (TH) that has been used to study the regulation of that enzyme.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluoro-L-phenylalanine is a substrate for tyrosine hydroxylase (TH) that has been used to study the regulation of that enzyme.
Solubility
Soluble in water and 0.5M HCl (50 mg/ml).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 1132-68-9 | |
| 183.182 | |
| XWHHYOYVRVGJJY-QMMMGPOBSA-N | |
| 716312 | |
| (2S)-2-amino-3-(4-fluorophenyl)propanoic acid |
| C9H10FNO2 | |
| MFCD00063064 | |
| 4-fluoro-l-phenylalanine, p-fluoro-l-phenylalanine, s-4-fluorophenylalanine, l-4-fluorophenylalanine, h-phe 4-f-oh, s-2-amino-3-4-fluorophenyl propanoic acid, unii-e2k2vdk6kk, l-phenylalanine, 4-fluoro, s---p-fluorophenylalanine, 2s-2-amino-3-4-fluorophenyl propanoic acid | |
| CHEBI:44909 | |
| C1=CC(=CC=C1CC(C(=O)O)N)F |
Specifications
| ∽240°C (decomposition) | |
| C9H10FNO2 | |
| 2416148 | |
| Soluble in water and 0.5M HCl (50mg/ml). | |
| C1=CC(=CC=C1CC(C(=O)O)N)F | |
| 250 mg | |
| 716312 | |
| −25° (c=1 in Water) | |
| ≥98% |
| 1132-68-9 | |
| MFCD00063064 | |
| 4-fluoro-l-phenylalanine, p-fluoro-l-phenylalanine, s-4-fluorophenylalanine, l-4-fluorophenylalanine, h-phe 4-f-oh, s-2-amino-3-4-fluorophenyl propanoic acid, unii-e2k2vdk6kk, l-phenylalanine, 4-fluoro, s---p-fluorophenylalanine, 2s-2-amino-3-4-fluorophenyl propanoic acid | |
| XWHHYOYVRVGJJY-QMMMGPOBSA-N | |
| (2S)-2-amino-3-(4-fluorophenyl)propanoic acid | |
| 183.182 | |
| CHEBI:44909 | |
| 183.18 | |
| 4-Fluoro-L-phenylalanine |
Safety and Handling
RTECSNumber : DW1765000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only