missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-L-2-phenylglycine 98.0+%, TCI America™

Catalog No. F08625G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08625G 5 g
F08621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08625G Supplier TCI America Supplier No. F08625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19883-57-9
Molecular Formula C8H8FNO2
Molecular Weight (g/mol) 169.16
MDL Number MFCD00213799
InChI Key JKFYKCYQEWQPTM-ZETCQYMHSA-N
Synonym s-4-fluorophenylglycine, s-2-amino-2-4-fluorophenyl acetic acid, 4-fluoro-l-alpha-phenylglycine, l-4-fluorophenylglycine, 4-fluoro-l-phenylglycine, l +-4-fluorophenylglycine, 2s-2-amino-2-4-fluorophenyl acetic acid, s-+-4-fluorophenyl glycine, p-fluorophenyl glycine, 4-fluorophenylglycine
PubChem CID 853015
IUPAC Name (2S)-2-azaniumyl-2-(4-fluorophenyl)acetate
SMILES [NH3+][C@H](C([O-])=O)C1=CC=C(F)C=C1

Specifications

Color White
Quantity 5 g
Formula Weight 169.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-L-2-phenylglycine