missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-DL-mandelic Acid 96.0+%, TCI America™

Catalog No. p-7129969
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09075G 5 g
F090725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09075G Supplier TCI America Supplier No. F09075G
Only null left

Chemical Identifiers

CAS 395-33-5
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.14
MDL Number MFCD00044402,MFCD20638902
InChI Key RWCMOQXHIDWDDJ-UHFFFAOYNA-N
Synonym 4-fluoromandelic acid, 2-4-fluorophenyl-2-hydroxyacetic acid, p-fluoro mandelic acid, 4-fluorophenyl hydroxy acetic acid, p-fluoromandelic acid, 4-fluoro mandelic acid, 4-fluoromandelicacid, 4-fluoro-alpha-hydroxybenzeneacetic acid, 4-fluoro-dl-mandelic acid, 4-fluoro hydroxy acetic acid
PubChem CID 98068
IUPAC Name 2-(4-fluorophenyl)-2-hydroxyacetic acid
SMILES OC(C(O)=O)C1=CC=C(F)C=C1

Specifications

Melting Point 137°C
Color White
Quantity 5 g
Formula Weight 170.14
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-DL-mandelic Acid