missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-D-2-phenylglycine 98.0+%, TCI America™

Catalog No. F08631G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08631G 1 g
F08635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08631G Supplier TCI America Supplier No. F08631G
Only null left

Chemical Identifiers

CAS 93939-74-3
Molecular Formula C8H8FNO2
Molecular Weight (g/mol) 169.155
MDL Number MFCD00042727
InChI Key JKFYKCYQEWQPTM-SSDOTTSWSA-N
Synonym r-4-fluorophenylglycine, r-2-amino-2-4-fluorophenyl acetic acid, d--4-fluorophenylglycine, 4-fluoro-d-phenylglycine, 4-fluoro-d-alpha-phenylglycine, d-4-fluorophenylglycine, r---4-fluorophenyl glycine, 4-fluoro-d-2-phenylglycine, 4-fluorophenylglycine, 4-fluoro-d-a-phenylglycine
PubChem CID 124515
IUPAC Name (2R)-2-amino-2-(4-fluorophenyl)acetic acid
SMILES C1=CC(=CC=C1C(C(=O)O)N)F

Specifications

Color White
Quantity 1 g
Formula Weight 169.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-D-2-phenylglycine