missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-alpha-toluenesulfonyl chloride, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 58°C to 61°C |
| Quantity | 1 g |
| UN Number | UN3261 |
| Sensitivity | Moisture sensitive |
| Formula Weight | 208.64 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Fluoro-alpha-toluenesulfonyl chloride |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only