missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-alpha-methylstyrene (stabilized with TBC) 98.0+%, TCI America™
Supplier: TCI America F025025ML
Specifications
| 4-Fluoro-alpha-methylstyrene (stabilized with TBC) | |
| 183°C | |
| MFCD00042297 | |
| 1993 | |
| VIXHMBLBLJSGIB-UHFFFAOYSA-N | |
| 1-fluoro-4-prop-1-en-2-ylbenzene | |
| 67690 | |
| ≥98.0% (GC) |
| 350-40-3 | |
| C9H9F | |
| 25 mL | |
| 4-fluoro-alpha-methylstyrene, 1-fluoro-4-1-methylethenyl benzene, 1-fluoro-4-prop-1-en-2-yl benzene, 2-4-fluorophenyl-1-propene, 2-4-fluorophenyl propene, benzene, 1-fluoro-4-1-methylethenyl, 4-fluoro-.alpha.-methylstyrene, 1-fluoro-4-isopropenyl benzene, 1-fluoro-4-isopropenylbenzene, p-fluoro-.alpha.-methylstyrene | |
| CC(=C)C1=CC=C(C=C1)F | |
| 136.169 | |
| 136.17 | |
| Liquid |
Chemical Identifiers
| 350-40-3 | |
| 136.169 | |
| VIXHMBLBLJSGIB-UHFFFAOYSA-N | |
| 67690 | |
| CC(=C)C1=CC=C(C=C1)F |
| C9H9F | |
| MFCD00042297 | |
| 4-fluoro-alpha-methylstyrene, 1-fluoro-4-1-methylethenyl benzene, 1-fluoro-4-prop-1-en-2-yl benzene, 2-4-fluorophenyl-1-propene, 2-4-fluorophenyl propene, benzene, 1-fluoro-4-1-methylethenyl, 4-fluoro-.alpha.-methylstyrene, 1-fluoro-4-isopropenyl benzene, 1-fluoro-4-isopropenylbenzene, p-fluoro-.alpha.-methylstyrene | |
| 1-fluoro-4-prop-1-en-2-ylbenzene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator