missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-alpha-methylstyrene, 95%

CAS: 350-40-3 | C9H9F | 136.169 g/mol

Supplier:  Thermo Scientific Chemicals A1414730

 View more versions of this product

Catalog No. AAA1414730

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Fluoro-alpha-methylstyrene
1.009
53°C (127°F)
1.511
250 g
2039696
VIXHMBLBLJSGIB-UHFFFAOYSA-N
1-fluoro-4-prop-1-en-2-ylbenzene
67690
95%
350-40-3
183°C
C9H9F
MFCD00042297
UN1993
4-fluoro-alpha-methylstyrene, 1-fluoro-4-1-methylethenyl benzene, 1-fluoro-4-prop-1-en-2-yl benzene, 2-4-fluorophenyl-1-propene, 2-4-fluorophenyl propene, benzene, 1-fluoro-4-1-methylethenyl, 4-fluoro-.alpha.-methylstyrene, 1-fluoro-4-isopropenyl benzene, 1-fluoro-4-isopropenylbenzene, p-fluoro-.alpha.-methylstyrene
CC(=C)C1=CC=C(C=C1)F
136.169
136.17

Chemical Identifiers

350-40-3
136.169
VIXHMBLBLJSGIB-UHFFFAOYSA-N
67690
CC(=C)C1=CC=C(C=C1)F
C9H9F
MFCD00042297
4-fluoro-alpha-methylstyrene, 1-fluoro-4-1-methylethenyl benzene, 1-fluoro-4-prop-1-en-2-yl benzene, 2-4-fluorophenyl-1-propene, 2-4-fluorophenyl propene, benzene, 1-fluoro-4-1-methylethenyl, 4-fluoro-.alpha.-methylstyrene, 1-fluoro-4-isopropenyl benzene, 1-fluoro-4-isopropenylbenzene, p-fluoro-.alpha.-methylstyrene
1-fluoro-4-prop-1-en-2-ylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 206-501-2

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only