missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-4'-(trans-4-propylcyclohexyl)biphenyl 98.0+%, TCI America™

Catalog No. F11031G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11031G 1 g
F11035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F11031G Supplier TCI America Supplier No. F11031G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87260-24-0
Molecular Formula C21H25F
Molecular Weight (g/mol) 296.429
MDL Number MFCD11053438
InChI Key DYBQDZYHNOGWIK-UHFFFAOYSA-N
Synonym 4-fluoro-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, trans-4-4-propylcyclohexyl-4'-fluorobiphenyl, 1-fluoro-4-4-4-propylcyclohexyl phenyl benzene, 4-fluoro-4'-1s,4r-4-propylcyclohexyl-1,1'-biphenyl, 4-4-propylcyclohexyl-4'-fluorobiphenyl, 4-fluoro-4'-trans-4-proptlcyclohexyl biphenyl, rans-4-4-propylcyclohexyl-4-fluorobiphenyl, trans-4-4-propylcyclohexyl-4-fluorobiphenyl, trans-4-fluoro-4'-4-n-propylcyclohexyl biphenyl, 1,1'-biphenyl, 4-fluoro-4'-4-propylcyclohexyl
PubChem CID 611152
IUPAC Name 1-fluoro-4-[4-(4-propylcyclohexyl)phenyl]benzene
SMILES CCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC=C(C=C3)F

Specifications

Melting Point 106°C
Color White
Quantity 1 g
Formula Weight 296.43
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-4′-(trans-4-propylcyclohexyl)biphenyl