accessibility menu, dialog, popup

UserName

CAS RN 87260-24-0

IUPAC Name: 4-fluoro-4'-(4-propylcyclohexyl)-1,1'-biphenyl
Synonyms: 4-fluoro-4'-(4-propylcyclohexyl)-1,1'-biphenyl
Molecular Weight (g/mol): 296.43
Molecular Formula: C21H25F
InChi Key: DYBQDZYHNOGWIK-UHFFFAOYSA-N
SMILES: CCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results