missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-4'-hydroxybiphenyl 98.0+%, TCI America™

Catalog No. F07045G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F07045G Supplier TCI America Supplier No. F07045G

Chemical Identifiers

CAS 324-94-7
Molecular Formula C12H9FO
Molecular Weight (g/mol) 188.201
MDL Number MFCD01830385
InChI Key QSJNKJGPJVOGPK-UHFFFAOYSA-N
Synonym 4'-fluoro-1,1'-biphenyl-4-ol, 4-fluoro-4'-hydroxybiphenyl, 4-hydroxy-4'-fluorobiphenyl, 4-4-fluorophenyl phenol, 4'-fluoro-biphenyl-4-ol, 4'-fluoro 1,1'-biphenyl-4-ol, 4-4'-fluorophenyl phenol, 4'-fluorobiphenyl-1-ol, 1,1'-biphenyl-4-ol, 4'-fluoro, pubchem9057
PubChem CID 345420
IUPAC Name 4-(4-fluorophenyl)phenol
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)F)O

Specifications

Melting Point 172°C
Color White-Yellow
Quantity 5 g
Formula Weight 188.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-4′-hydroxybiphenyl