missing translation for 'onlineSavingsMsg'
Learn More

[4-Fluoro-3-(trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate 97.0+%, TCI America™

Catalog No. F11111G
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11111G 1 g
F1111200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F11111G Supplier TCI America Supplier No. F11111G

Chemical Identifiers

CAS 1868173-29-8
Molecular Formula C23H21F4IO6S
Molecular Weight (g/mol) 628.373
InChI Key JRLOQNMMXUNDFI-UHFFFAOYSA-M
Synonym [4-Fluoro-3-(trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium Tosylate
PubChem CID 131697918
IUPAC Name [4-fluoro-3-(trifluoromethyl)phenyl]-(2,4,6-trimethoxyphenyl)iodanium;4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)[O-].COC1=CC(=C(C(=C1)OC)[I+]C2=CC(=C(C=C2)F)C(F)(F)F)OC

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 628.37
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material [4-Fluoro-3-(trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate
TSCA No
Recommended Storage Refrigerator