missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(trifluoromethyl)benzaldehyde 97.0+%, TCI America™

Catalog No. F05451G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05451G 1 g
F05455G 5 g
F054525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F05451G Supplier TCI America Supplier No. F05451G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67515-60-0
Molecular Formula C8H4F4O
Molecular Weight (g/mol) 192.113
InChI Key BIUDHHGROGJSHN-UHFFFAOYSA-N
Synonym 4-fluoro-3-trifluoromethyl benzaldehyde, 3-trifluoromethyl-4-fluorobenzaldehyde, benzaldehyde, 4-fluoro-3-trifluoromethyl, alpha,alpha,alpha,4-tetrafluoro-m-tolualdehyde, pubchem1462, acmc-209nzm, ksc359a9l, 4-fluoro-3-trifluoromethyl
PubChem CID 522269
IUPAC Name 4-fluoro-3-(trifluoromethyl)benzaldehyde
SMILES C1=CC(=C(C=C1C=O)C(F)(F)F)F

Specifications

Color Yellow
Boiling Point 60°C
Quantity 1 g
Formula Weight 192.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzaldehyde