missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitrophenyl Methyl Sulfone 98.0+%, TCI America™

Catalog No. F08871G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08871G 1 g
F08875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08871G Supplier TCI America Supplier No. F08871G

Chemical Identifiers

CAS 453-72-5
Molecular Formula C7H6FNO4S
Molecular Weight (g/mol) 219.19
MDL Number MFCD00025066
InChI Key OUSNDSFSTBZESM-UHFFFAOYSA-N
Synonym 4-methylsulfonyl-2-nitrofluorobenzene, 1-fluoro-4-methylsulfonyl-2-nitrobenzene, 1-fluoro-4-methanesulfonyl-2-nitrobenzene, 2-fluoro-5-methyl sulfonyl nitrobenzene, 2-fluoro-5-methylsulphonylnitrobenzene, 4-fluoro-3-nitrophenyl methyl sulfone, benzene,1-fluoro-4-methylsulfonyl-2-nitro, 2-fluoro-5-methylsulfonylnitrobenzene, 2-fluoro-5-methylsulphonyl nitrobenzene, 1-fluoro-4-methylsulfonyl-2-nitro-benzene
PubChem CID 313137
IUPAC Name 1-fluoro-4-methanesulfonyl-2-nitrobenzene
SMILES CS(=O)(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O

Specifications

Melting Point 158°C
Color Yellow
Quantity 1 g
Formula Weight 219.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-3-nitrophenyl Methyl Sulfone
TSCA No
Recommended Storage Refrigerator