missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitrobenzonitrile 98.0+%, TCI America™

Catalog No. p-7130041
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10405G 5 g
F104025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10405G Supplier TCI America Supplier No. F10405G
Only null left

Chemical Identifiers

CAS 1009-35-4
Molecular Formula C7H3FN2O2
Molecular Weight (g/mol) 166.111
MDL Number MFCD01632197
InChI Key LKOWKPGBAZVHOF-UHFFFAOYSA-N
Synonym 3-nitro-4-fluorobenzonitrile, 4-fluoro-3-nitro-benzonitrile, 5-cyano-2-fluoronitrobenzene, 4-cyano-1-fluoro-2-nitrobenzene, benzonitrile, 4-fluoro-3-nitro, 4-fluoro-3-nitrobenzenecarbonitrile, 5-cyano-5-fluoro-1-nitrobenzene, pubchem4796, acmc-1c2vd, 4-fluro-3-nitrobenzonitrile
PubChem CID 2774654
IUPAC Name 4-fluoro-3-nitrobenzonitrile
SMILES C1=CC(=C(C=C1C#N)[N+](=O)[O-])F

Specifications

Melting Point 93°C
Color Yellow
Quantity 5 g
UN Number 3439
Formula Weight 166.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-3-nitrobenzonitrile