missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitrobenzonitrile, 97%

Catalog No. p-7031348
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3239103 1 g
AAH3239106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3239103 Supplier Thermo Scientific Chemicals Supplier No. H3239103
Only null left

CAS: 1009-35-4 | C7H3FN2O2 | 166.111 g/mol

4-Fluoro-3-nitrobenzonitrile, is used as a pharmaceutical intermediate. It is also used as a synthetic and fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-3-nitrobenzonitrile, is used as a pharmaceutical intermediate. It is also used as a synthetic and fine chemical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Chemical Identifiers

CAS 1009-35-4
Molecular Formula C7H3FN2O2
Molecular Weight (g/mol) 166.111
MDL Number MFCD01632197
InChI Key LKOWKPGBAZVHOF-UHFFFAOYSA-N
Synonym 3-nitro-4-fluorobenzonitrile, 4-fluoro-3-nitro-benzonitrile, 5-cyano-2-fluoronitrobenzene, 4-cyano-1-fluoro-2-nitrobenzene, benzonitrile, 4-fluoro-3-nitro, 4-fluoro-3-nitrobenzenecarbonitrile, 5-cyano-5-fluoro-1-nitrobenzene, pubchem4796, acmc-1c2vd, 4-fluro-3-nitrobenzonitrile
PubChem CID 2774654
IUPAC Name 4-fluoro-3-nitrobenzonitrile
SMILES C1=CC(=C(C=C1C#N)[N+](=O)[O-])F

Specifications

Melting Point 90°C to 94°C
Quantity 1 g
UN Number UN3439
Solubility Information Insoluble in water.
Formula Weight 166.11
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-nitrobenzonitrile
Hazard Category H302+H332-H311-H315-H319
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only