missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-3-nitrobenzoic Acid 98.0+%, TCI America™
$72.70 - $231.14
Chemical Identifiers
| CAS | 453-71-4 |
|---|---|
| Molecular Formula | C7H4FNO4 |
| Molecular Weight (g/mol) | 185.11 |
| MDL Number | MFCD00007058 |
| InChI Key | BOJWTAQWPVBIPG-UHFFFAOYSA-N |
| Synonym | 4-fluoro-3-nitrobenzoic acid, 3-nitro-4-fluorobenzoic acid, p-fluoro-3-nitrobenzoic acid, 4-fluoro-3-nitrobenzoicacid, 5-carboxy-2-fluoronitrobenzene, benzoic acid, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzoic acid, pubchem1306, acmc-1aqvm, 3-nitro-4-flurobenzoic acid |
| PubChem CID | 67987 |
| IUPAC Name | 4-fluoro-3-nitrobenzoic acid |
| SMILES | OC(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O |
Chemical Identifiers
| 453-71-4 | |
| 185.11 | |
| BOJWTAQWPVBIPG-UHFFFAOYSA-N | |
| 67987 | |
| OC(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O |
| C7H4FNO4 | |
| MFCD00007058 | |
| 4-fluoro-3-nitrobenzoic acid, 3-nitro-4-fluorobenzoic acid, p-fluoro-3-nitrobenzoic acid, 4-fluoro-3-nitrobenzoicacid, 5-carboxy-2-fluoronitrobenzene, benzoic acid, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzoic acid, pubchem1306, acmc-1aqvm, 3-nitro-4-flurobenzoic acid | |
| 4-fluoro-3-nitrobenzoic acid |
Specifications
| 453-71-4 | |
| Yellow | |
| MFCD00007058 | |
| 4-fluoro-3-nitrobenzoic acid, 3-nitro-4-fluorobenzoic acid, p-fluoro-3-nitrobenzoic acid, 4-fluoro-3-nitrobenzoicacid, 5-carboxy-2-fluoronitrobenzene, benzoic acid, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzoic acid, pubchem1306, acmc-1aqvm, 3-nitro-4-flurobenzoic acid | |
| OC(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O | |
| 185.11 | |
| 185.11 | |
| Crystalline Powder |
| 121°C | |
| C7H4FNO4 | |
| 5 g | |
| BOJWTAQWPVBIPG-UHFFFAOYSA-N | |
| 4-fluoro-3-nitrobenzoic acid | |
| 67987 | |
| ≥98.0% (GC,T) | |
| 4-Fluoro-3-nitrobenzoic Acid |
Safety and Handling
TSCA : No