missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. F03995G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03995G 5 g
F039925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03995G Supplier TCI America Supplier No. F03995G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 453-71-4
Molecular Formula C7H4FNO4
Molecular Weight (g/mol) 185.11
MDL Number MFCD00007058
InChI Key BOJWTAQWPVBIPG-UHFFFAOYSA-N
Synonym 4-fluoro-3-nitrobenzoic acid, 3-nitro-4-fluorobenzoic acid, p-fluoro-3-nitrobenzoic acid, 4-fluoro-3-nitrobenzoicacid, 5-carboxy-2-fluoronitrobenzene, benzoic acid, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzoic acid, pubchem1306, acmc-1aqvm, 3-nitro-4-flurobenzoic acid
PubChem CID 67987
IUPAC Name 4-fluoro-3-nitrobenzoic acid
SMILES OC(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O

Specifications

Melting Point 121°C
Color Yellow
Quantity 5 g
Formula Weight 185.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-3-nitrobenzoic Acid