Learn More
4-Fluoro-3-nitrobenzoic acid, 98%
CAS: 453-71-4 | C7H4FNO4 | 185.11 g/mol
Supplier: Thermo Scientific Chemicals A1080014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Fluoro-3-nitrobenzoic acid | |
| 121°C to 126°C | |
| C7H4FNO4 | |
| 25 g | |
| 4-fluoro-3-nitrobenzoic acid, 3-nitro-4-fluorobenzoic acid, p-fluoro-3-nitrobenzoic acid, 4-fluoro-3-nitrobenzoicacid, 5-carboxy-2-fluoronitrobenzene, benzoic acid, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzoic acid, pubchem1306, acmc-1aqvm, 3-nitro-4-flurobenzoic acid | |
| OC(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O | |
| 185.11 | |
| 185.11 |
| 453-71-4 | |
| 92°C (15 mmHg) | |
| MFCD00007058 | |
| 2107562 | |
| BOJWTAQWPVBIPG-UHFFFAOYSA-N | |
| 4-fluoro-3-nitrobenzoic acid | |
| 67987 | |
| 98% |
Chemical Identifiers
| 453-71-4 | |
| 185.11 | |
| BOJWTAQWPVBIPG-UHFFFAOYSA-N | |
| 67987 | |
| OC(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O |
| C7H4FNO4 | |
| MFCD00007058 | |
| 4-fluoro-3-nitrobenzoic acid, 3-nitro-4-fluorobenzoic acid, p-fluoro-3-nitrobenzoic acid, 4-fluoro-3-nitrobenzoicacid, 5-carboxy-2-fluoronitrobenzene, benzoic acid, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzoic acid, pubchem1306, acmc-1aqvm, 3-nitro-4-flurobenzoic acid | |
| 4-fluoro-3-nitrobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 207-221-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only