missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitrobenzoic acid, 98%

Catalog No. AAA1080022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1080022 100 g
AAA1080006 5 g
AAA1080014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1080022 Supplier Thermo Scientific Chemicals Supplier No. A1080022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 453-71-4 | C7H4FNO4 | 185.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 453-71-4
Molecular Formula C7H4FNO4
Molecular Weight (g/mol) 185.11
MDL Number MFCD00007058
InChI Key BOJWTAQWPVBIPG-UHFFFAOYSA-N
Synonym 4-fluoro-3-nitrobenzoic acid, 3-nitro-4-fluorobenzoic acid, p-fluoro-3-nitrobenzoic acid, 4-fluoro-3-nitrobenzoicacid, 5-carboxy-2-fluoronitrobenzene, benzoic acid, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzoic acid, pubchem1306, acmc-1aqvm, 3-nitro-4-flurobenzoic acid
PubChem CID 67987
IUPAC Name 4-fluoro-3-nitrobenzoic acid
SMILES OC(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O

Specifications

Melting Point 121°C to 126°C
Boiling Point 92°C (15 mmHg)
Quantity 100 g
Beilstein 2107562
Formula Weight 185.11
Percent Purity 98%
Chemical Name or Material 4-Fluoro-3-nitrobenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 207-221-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only