missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitroanisole, 95%

Catalog No. AAH6155406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6155406 5 g
AAH6155403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6155406 Supplier Thermo Scientific Chemicals Supplier No. H6155406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 61324-93-4 | C7H6FNO3 | 171.127 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61324-93-4
Molecular Formula C7H6FNO3
Molecular Weight (g/mol) 171.127
MDL Number MFCD11848442
InChI Key ZRIKJXDEJYMBEJ-UHFFFAOYSA-N
Synonym 4-fluoro-3-nitroanisole, benzene, 1-fluoro-4-methoxy-2-nitro, 3-fluoro-4-nitrobenzene,-1-methoxy, ksc613a5f, 2-fluoro-5-methoxynitrobenzene, 2-fluoro-5-methoxy-nitrobenzene, #
PubChem CID 565653
IUPAC Name 1-fluoro-4-methoxy-2-nitrobenzene
SMILES COC1=CC(=C(C=C1)F)[N+](=O)[O-]

Specifications

Quantity 5 g
Formula Weight 171.13
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Fluoro-3-nitroanisole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only