missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitroaniline, 98%

Catalog No. p-7022543
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1537114 25 g
AAA1537122 100 g
AAA1537136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1537114 Supplier Thermo Scientific Chemicals Supplier No. A1537114
Only null left

CAS: 364-76-1 | C6H5FN2O2 | 156.12 g/mol

4-Fluoro-3-nitroaniline is used in the preparation of commercial hair dyes. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-3-nitroaniline is used in the preparation of commercial hair dyes. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Insoluble in water.

Notes
Incompatible with Oxidizing agents, Acids, Acid chlorides, Acid anhydrides.

Chemical Identifiers

CAS 364-76-1
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.12
MDL Number MFCD00007833
InChI Key LLIOADBCFIXIEU-UHFFFAOYSA-N
Synonym 3-nitro-4-fluoroaniline, 4-fluoro-3-nitrobenzenamine, benzenamine, 4-fluoro-3-nitro, aniline, 4-fluoro-3-nitro, 4-fluoro-3-nitro-aniline, 5-amino-2-fluoronitrobenzene, 4-fluoro-3-nitro-phenylamine, 3-nitro-4-fluoroanailine, 3-nitro-4-fluoro aniline, 4-fluoro-3-nitro aniline
PubChem CID 67768
ChEBI CHEBI:48642
IUPAC Name 4-fluoro-3-nitroaniline
SMILES NC1=CC=C(F)C(=C1)[N+]([O-])=O

Specifications

Melting Point 94°C to 98°C
Flash Point 91°C (195°F)
Quantity 25 g
Beilstein 2210199
Solubility Information Insoluble in water.
Formula Weight 156.12
Percent Purity 98%
Chemical Name or Material 4-Fluoro-3-nitroaniline
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 206-665-5
RTECSNumber BY1700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only