missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-3-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$130.92 - $130.92
Specifications
| CAS | 139911-27-6 |
|---|---|
| Melting Point | 217°C |
| Color | White-Yellow |
| Molecular Formula | C7H8BFO2 |
| MDL Number | MFCD01863527 |
Specifications
| 139911-27-6 | |
| White-Yellow | |
| MFCD01863527 | |
| (4-fluoro-3-methylphenyl)boronic acid | |
| 153.95 |
| 217°C | |
| C7H8BFO2 | |
| JCIJCHSRVPSOML-UHFFFAOYSA-N | |
| 2774580 | |
| Crystalline Powder |