missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methylphenylboronic acid, 98%

Catalog No. p-4228699
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC435330050 5 g Glass bottle
AC435330250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC435330050 Supplier Thermo Scientific Chemicals Supplier No. 435330050

CAS: 139911-27-6 | C7H8BFO2 | 153.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 139911-27-6
Molecular Formula C7H8BFO2
Molecular Weight (g/mol) 153.95
MDL Number MFCD01863527
InChI Key JCIJCHSRVPSOML-UHFFFAOYSA-N
Synonym 3-methyl-4-fluorophenylboronic acid, 4-fluoro-3-methylphenyl boronic acid, 4-fluoro-3-methylbenzeneboronic acid, 4-fluoro-m-tolylboronic acid, 4-fluoro-3-methylphenylboronicacid, 5-borono-2-fluorotoluene, boronic acid, 4-fluoro-3-methylphenyl, pubchem5146, acmc-209cjo
PubChem CID 2774580
IUPAC Name (4-fluoro-3-methylphenyl)boronic acid
SMILES CC1=CC(=CC=C1F)B(O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 215.4°C to 217.3°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 153.95
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-3-methylphenylboronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only