missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methylbenzonitrile 98.0+%, TCI America™

Catalog No. p-7130013
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09865G 5 g
F098625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09865G Supplier TCI America Supplier No. F09865G
Only null left

Chemical Identifiers

CAS 185147-08-4
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.14
MDL Number MFCD03788510
InChI Key ZMEAHKIIWJDJFT-UHFFFAOYSA-N
Synonym 3-methyl-4-fluorobenzonitrile, 3-methyl-4-fluoro benzonitrile, benzonitrile, 4-fluoro-3-methyl, 4-fluoro-3-methyl-benzonitrile, 2-fluoro-5-cyanotoluene, 5-cyano-2-fluorotoluene, 4-fluoro-3-methylbenzenecarbonitrile, pubchem1553, 4-fluoro-m-tolunitrile, acmc-209emo
PubChem CID 2779180
IUPAC Name 4-fluoro-3-methylbenzonitrile
SMILES CC1=CC(=CC=C1F)C#N

Specifications

Melting Point 56°C
Color White-Yellow
Quantity 5 g
Formula Weight 135.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-3-methylbenzonitrile