missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methylaniline 98.0+%, TCI America™

Catalog No. p-7129731
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05275G 5 g
F052725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05275G Supplier TCI America Supplier No. F05275G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 452-69-7
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00025294
InChI Key NYMDPDNETOLVBS-UHFFFAOYSA-N
Synonym 2-fluoro-5-aminotoluene, 4-fluoro-m-toluidine, 5-amino-2-fluorotoluene, 4-fluoro-3-methylbenzenamine, 4-fluoro-3-methylphenylamine, benzenamine, 4-fluoro-3-methyl, 4-fluoro-3-methyl aniline, 4-fluoro-3-methyl-aniline, 4-fluor-3-methylanilin, pubchem1513
PubChem CID 67981
IUPAC Name 4-fluoro-3-methylaniline
SMILES CC1=CC(N)=CC=C1F

Specifications

Melting Point 36°C
Color Gray-White
Boiling Point 86°C
Quantity 5 g
Formula Weight 125.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-3-methylaniline