missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoro-3'-methylacetophenone 98.0+%, TCI America™

Catalog No. p-7130066
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10971G 1 g
F10975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10971G Supplier TCI America Supplier No. F10971G
Only null left

Chemical Identifiers

CAS 369-32-4
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.17
MDL Number MFCD04972879
InChI Key SMSVMBMJEYTUOZ-UHFFFAOYSA-N
PubChem CID 289328
IUPAC Name 1-(4-fluoro-3-methylphenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(F)C(C)=C1

Specifications

Boiling Point 215°C
Quantity 1 g
Synonym 1-4-fluoro-3-methylphenyl ethanone, 4-fluoro-3-methylacetophenone, 4'-fluoro-3'-methylacetophenone, 1-4-fluoro-3-methylphenyl ethan-1-one, 3-methyl-4-fluoroacetophenone, 1-acetyl-4-fluoro-3-methylbenzene, ethanone,1-4-fluoro-3-methylphenyl, ethanone, 1-4-fluoro-3-methylphenyl, pubchem19762, 4-fluor-3-methylacetophenone
Formula Weight 152.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Fluoro-3′-methylacetophenone