accessibility menu, dialog, popup

UserName

H3COFH3C

CAS RN 369-32-4

IUPAC Name: 1-(4-fluoro-3-methylphenyl)ethan-1-one
Synonyms: 1-(4-fluoro-3-methylphenyl)ethanone
Molecular Weight (g/mol): 152.17
Molecular Formula: C9H9FO
InChi Key: SMSVMBMJEYTUOZ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(F)C(C)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results