missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methoxybenzoic Acid 98.0+%, TCI America™

Catalog No. p-7129831
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06811G 1 g
F06815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06811G Supplier TCI America Supplier No. F06811G
Only null left

Chemical Identifiers

CAS 82846-18-2
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD00272134
InChI Key LWGCZCMLPRMKIZ-UHFFFAOYSA-N
Synonym 3-methoxy-4-fluorobenzoic acid, 4-fluoro-m-anisic acid, benzoic acid, 4-fluoro-3-methoxy, 4-fluoro-3-methoxybenzoicacid, 4-fluoro-3-methoxy-benzoic acid, 4-fluoranyl-3-methoxy-benzoic acid, pubchem1394, acmc-209pqm, ksc495c0r, rarechem al bo 0973
PubChem CID 598436
IUPAC Name 4-fluoro-3-methoxybenzoic acid
SMILES COC1=C(C=CC(=C1)C(=O)O)F

Specifications

Melting Point 207°C
Color White-Yellow
Quantity 1 g
Formula Weight 170.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-3-methoxybenzoic Acid