Learn More
4-Fluoro-3-methoxybenzoic acid, 95%, Thermo Scientific Chemicals
$301.86 - $301.86
Chemical Identifiers
| CAS | 82846-18-2 |
|---|---|
| Molecular Formula | C8H7FO3 |
| Molecular Weight (g/mol) | 170.139 |
| MDL Number | MFCD00272134 |
| InChI Key | LWGCZCMLPRMKIZ-UHFFFAOYSA-N |
| Synonym | 3-methoxy-4-fluorobenzoic acid, 4-fluoro-m-anisic acid, benzoic acid, 4-fluoro-3-methoxy, 4-fluoro-3-methoxybenzoicacid, 4-fluoro-3-methoxy-benzoic acid, 4-fluoranyl-3-methoxy-benzoic acid, pubchem1394, acmc-209pqm, ksc495c0r, rarechem al bo 0973 |
| PubChem CID | 598436 |
| IUPAC Name | 4-fluoro-3-methoxybenzoic acid |
| SMILES | COC1=C(C=CC(=C1)C(=O)O)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1967306
|
Thermo Scientific Chemicals
L1967306 |
5 g |
Each for $301.86
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 82846-18-2 | |
| 170.139 | |
| LWGCZCMLPRMKIZ-UHFFFAOYSA-N | |
| 598436 | |
| COC1=C(C=CC(=C1)C(=O)O)F |
| C8H7FO3 | |
| MFCD00272134 | |
| 3-methoxy-4-fluorobenzoic acid, 4-fluoro-m-anisic acid, benzoic acid, 4-fluoro-3-methoxy, 4-fluoro-3-methoxybenzoicacid, 4-fluoro-3-methoxy-benzoic acid, 4-fluoranyl-3-methoxy-benzoic acid, pubchem1394, acmc-209pqm, ksc495c0r, rarechem al bo 0973 | |
| 4-fluoro-3-methoxybenzoic acid |
Specifications
| 82846-18-2 | |
| C8H7FO3 | |
| 5 g | |
| 3-methoxy-4-fluorobenzoic acid, 4-fluoro-m-anisic acid, benzoic acid, 4-fluoro-3-methoxy, 4-fluoro-3-methoxybenzoicacid, 4-fluoro-3-methoxy-benzoic acid, 4-fluoranyl-3-methoxy-benzoic acid, pubchem1394, acmc-209pqm, ksc495c0r, rarechem al bo 0973 | |
| COC1=C(C=CC(=C1)C(=O)O)F | |
| 170.139 | |
| 170.14 | |
| 4-Fluoro-3-methoxybenzoic acid |
| 206°C to 208°C | |
| MFCD00272134 | |
| 4178071 | |
| LWGCZCMLPRMKIZ-UHFFFAOYSA-N | |
| 4-fluoro-3-methoxybenzoic acid | |
| 598436 | |
| 95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only