missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(hydroxymethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7130062
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1090200MG 200 mg
F10901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F1090200MG Supplier TCI America Supplier No. F1090200MG

Chemical Identifiers

CAS 481681-02-1
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.95
MDL Number MFCD08235075
InChI Key PWMOQHMTXJYUGE-UHFFFAOYSA-N
Synonym 4-fluoro-3-hydroxymethyl phenyl boronic acid, 4-fluoro-3-hydroxymethyl phenylboronic acid, 4-fluoro-3-hydroxymethyl benzeneboronic acid, 4-fluoro-3-hydroxymethylphenylboronic acid, boronic acid, 4-fluoro-3-hydroxymethyl phenyl, boronic acid,b-4-fluoro-3-hydroxymethyl phenyl, pubchem23767, acmc-209kcj, ksc496e1t
PubChem CID 21865588
IUPAC Name [4-fluoro-3-(hydroxymethyl)phenyl]boronic acid
SMILES OCC1=C(F)C=CC(=C1)B(O)O

Specifications

Melting Point 126°C
Color White-Yellow
Quantity 200 mg
Formula Weight 169.95
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-3-(hydroxymethyl)phenylboronic Acid (contains varying amounts of Anhydride)